Description
- One-part flowable, RTV silicone dispersed in xylene
- Cures at room temperature within 24 hours upon exposure to atmospheric moisture
- Non-corrosive oxime cure system
- Electrically insulating
Applications
- Conformal coating for rigid and flexible printed circuit boards (PCBs).
- Protects from moisture ingress, vibration, shock, dust, chemicals, and other environmental hazards
- Low viscosity ideal for use as sprayable or dip coatings
- Suitable for dip casting into thin films
Product Details
Property | Average Result |
---|---|
Viscosity | 3,400 cP (3,400 mPa*s) |
Non Volatile Content | 60 % |
Appearance | Translucent |
Tack Free Time | 20 minutes |
Cure | 6 days / RTV |
Durometer | 40 Type A |
Elongation | 330 % |
Tensile | 745 psi (5.14 MPa) |
![](/m-nusil.avantorsciences.com/en_US/images/satellite-in-space-375x235.jpg)
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.