Description
- Dimethylsiloxane polymer, hydride terminated
Applications
- Commonly used for reacting with vinyl functional groups to form crosslinks in silicone systems to form adhesives, sealants, rubbers and gels.
Product Details
Property | Average Result |
---|---|
Refractive Index | |
Viscosity | |
Mmol | 0.16 H/g |
Appearance | |
Volatile Content |