Description
- Specially formulated primer designed for use with platinum-cured systems on polycarbonates
- One-component primer supplied in VM&P Naphtha requires no mixing
- Air-drying
- Convenient container sizes produce less waste
Applications
- Improves the adhesion of addition-cured systems to polycarbonates
Product Details
Property | Average Result |
---|---|
Non Volatile Content | |
Specific Gravity | |
Appearance | |
Cure System | |
Comment |