Description
- Specially formulated primer designed for use with condensation cured systems where conventional silicone primers are insufficient
- One-component primer supplied in VM&P Naphtha requires no mixing
- Air-drying
- Convenient container sizes produce less waste
Applications
- Improves the adhesion of one and two-part condensation cured systems to various substrates including: metals, glass ceramics, some composites, rigid plastics , and wood
Product Details
Property | Average Result |
---|---|
Non Volatile Content | |
Specific Gravity | |
Appearance | |
Cure System | |
Comment |