Description
- Specially formulated primer designed for use with one and two-part RTV systems where conventional silicone primers are insufficient
- Translucent to preserve visibility with translucent products
- One-component primers supplied in VM&P Naphtha require no mixing
- Air-drying
- Convenient container sizes produce less waste
Applications
- Improves the adhesion of one and two-part RTV systems to various substrates including: metals, glass, ceramics, rigid plastics, and wood
Product Details
Property | Average Result |
---|---|
Non Volatile Content | |
Specific Gravity | |
Appearance | |
Cure System | |
Lap Shear | 315 psi (2.17 MPa) |
Comment |