SFM5-2350

FLAME RETARDANT, RTV SILICONE FOAM

Description

  • Yields a medium density, silicone foam when catalyzed
  • Two Parts mix in a convenient 1:1 ratio with a black Part A and white Part B to ensure homogeneous mixtures

Applications

  • As a flame resistant seal
  • In shock and vibration dampening situations requiring a lightweight, flexible foam with excellent thermal insulation and radiation resistance
  • Useful where stability at higher and lower temperatures is required
  • For intricate back-filling due to a low viscosity and longer work time

Product Details

Property Average Result
Viscosity
Cure System
Foam Density 25 lbs/ft3 (0.4 g/cm3)
Appearance
Cure
Dielectric Strength
Mix Ratio
Specific Gravity
Volume Resistivity
Work Time 23 minutes
Comment

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.