Description
- One-part RTV silicone elastomer dispersed in tert butyl acetate
- 100 mol% fluorosilicone
Applications
- For coating, sealing and bonding applications requiring solvent and/or fuel resistance
- Designed for spraying, dip casting, and RTV curing of thin, elastomeric films
- For use as a coating to minimize ice adhesion
- Bonds aggressively to most surfaces
- Slight addition of heat will accelerate cure
Product Details
Property | Average Result |
---|---|
Viscosity | |
Non Volatile Content | |
Appearance | |
Cure | |
Cure System | |
Durometer | |
Elongation | |
Specific Gravity | |
Tear | |
Tensile | |
Comment |