Description
- Two-part, white, thermally conductive silicone
- Cure may be heat accelerated
- 15:1 Mix Ratio (Part A: Part B)
Applications
- To provide heat transfer between electrical/electronic components and their heat sinks
- Use to adhere integrated circuit substrates, base plates, heat sinks or where grooves or other configurations require a non-flowable to limited flow material
Product Details
Property | Average Result |
---|---|
Thermal Conductivity | 1.46 W/(mK) |
Cure System | |
Tensile | |
Appearance | |
Cure | |
Dielectric Strength | |
Durometer | |
Elongation | |
Flow Rate | 0.3 inches (7.62 mm) |
Mix Ratio | |
Operating Temperature | |
Specific Gravity | |
Work Time | 3 hours |
Comment |