Description
- Solvent-free, low viscosity, clear, two-part silicone elastomer
- Accelerated cure rate with heat
- Based on a dimethyl diphenyl silicone copolymer with a broad service temperature range
- 10:1 Mix Ratio (Part A:Part B)
Applications
- For environmental protection in potting, encapsulating and coating applications requiring radiation resistance, low temperature flexibility, oxidation and thermal stability
- Low viscosity allows for easy flow around complex parts, providing excellent shock and electrical insulation resistance
- Formulated for extreme temperature applications
- For applications requiring an operating temperature range of -115 to 260°C (–175 to 500°F)
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Cure | |
Mix Ratio | |
Operating Temperature | |
Refractive Index | |
Specific Gravity | |
Comment |