Description
- Two-part, black, electrically conductive RTV silicone
- Based on a methyl silicone polymer with a broad temperature range
- 10:1 Mix Ratio (Base: Curing Agent)
Applications
- For RFI and EMI shielding for electrical and space applications
- Provides protection from humidity, radiation, thermal stress and mechanical stress
- Well suited for formed in place conductive gaskets
- Use to adhere covers on housings or where grooves or other configurations require a flowable to limited flow material with a low viscosity
- For applications requiring an operating temperature range of -65°C to 200°C (-85°F to 392°F)
Product Details
Property | Average Result |
---|---|
Cure System | |
Durometer | |
Appearance | |
Cure | |
Elongation | |
Mix Ratio | |
Operating Temperature | |
Specific Gravity | |
Tear | |
Tensile | |
Volume Resistivity | |
Work Time | 15 hours |
Comment |