Description
- Two-part, optically clear, solvent free, low viscosity silicone
- Cures at room temperature or rapidly with heat
- Offers good physical and electrical stability across a broad range of temperatures
- 10:1 Mix Ratio (Part A:Part B)
Applications
- To provide protection of electronic components and assemblies against shock, vibration, moisture, ozone, dust, chemicals and other environmental hazards by potting or encapsulating
- For photonics applications
- For applications requiring an operating temperature range of -65 to 240°C (-85 to 465°F)
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Cure | |
Dielectric Strength | |
Mix Ratio | |
Operating Temperature | |
Refractive Index | |
Specific Gravity | |
Tear | |
Volume Resistivity | |
Work Time | 4 hours |
Comment |