R-2615-3
RED POTTING AND ENCAPSULATING SILICONE ELASTOMER
Description
  • Two-part, red, solvent free, low viscosity silicone
  • Offers good physical and electrical stability across a broad range of temperatures
  • Convenient 1:1 mix ratio
  • 1:1 Mix Ratio (Part A:Part B)
Applications
  • To provide protection of electronic components and assemblies against shock, vibration, moisture, ozone, dust, chemicals and other environmental hazards by potting or encapsulating
  • For applications requiring an operating temperature range of –65 to 240°C (-85 to 465°F)
Product Details
Property Average Result
Viscosity
Durometer
Tensile
Elongation
Appearance
Cure
Dielectric Strength
Specific Gravity
Work Time 2 hours