Description
- Two part, Clear, 10:1 Mix ratio (A:B)
- Pourable and self-leveling
- Cures at room temperature
- REACh and RoHS compliant
- Tested per UL-94 and passed V-0
Applications
- Good optical clarity allows for photonic coupling and visual inspection during component assembly
- Low viscosity allows the potting of complex geometries without the entrapment of air
- Superior UV and weathering resistance for protection of outdoor electronic components
- Applications include: potting for optical sensors, components and various electronic devices
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Work Time | 2 hours |