R-2560
WIDE TEMPERATURE RANGE, RTV SILICONE ELASTOMER (RED)
Description
  • Two-part, pourable, red RTV silicone
  • 100:0.5 Mix Ratio (Base: Curing Agent)
  • Based on a dimethyl diphenyl silicone copolymer with a broad temperature range
  • Fast cure
Applications
  • For potting, sealing and bonding applications
  • For thermal insulating as a firewall sealant or as an ablative coating in missile, rockets, aircraft and launch equipment
  • For applications in extreme temperatures
  • For applications requiring an operating temperature range of –115°C to 260°C (-175°F to 500°F)
Product Details
Property Average Result
Cure System
Viscosity
Lap Shear 385 psi (2.66 MPa)
Durometer
Appearance
Cure
Elongation
Mix Ratio
Non Volatile Content
Operating Temperature
Specific Gravity
Tack Free Time
Tensile
Work Time 1 minutes
Comment