Description
- Two-part, pourable, translucent RTV silicone
- Based on a dimethyl diphenyl silicone copolymer with a broad temperature range
- 100: 0.5 Mix Ratio (Base: Curing Agent)
Applications
- As a heat shielding material for potting, sealing and bonding applications
- For ablative coatings in aerospace applications that require a material that pours
- For applications in extreme temperatures
- For applications requiring an operating temperature range of -115 to 260°C (-178 to 500°F)
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Appearance | |
Cure | |
Elongation | |
Mix Ratio | |
Non Volatile Content | |
Operating Temperature | |
Refractive Index | |
Specific Gravity | |
Tear | |
Tensile | |
Work Time | 6 hours |
Comment |