Description
- Two part, Clear, 1:1 Mix ratio (A:B)
- Pourable and self-leveling
- Cures at room temperature
- REACh and RoHS compliant
- Tested per UL-94 and passed V-1
Applications
- For potting, encapsulating and coating of modules, relays, and a variety of AC/DC converters including high power and planar packages
- Provides protection from moisture, dust and contaminants
- Increases resistance to shock and vibration
- Low viscosity allows the potting of complex geometries without the entrapment of air
- Pot life is compatible with processes that require dispensing or coating with Part A and B combined
- Dielectric properties compatible with many devices and increase breakdown voltage of device
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Cte | |
Cure | |
Dielectric Strength | |
Ionic Content Cl | <5 ppm |
Ionic Content K | <3 ppm |
Ionic Content Na | <1 ppm |
Mix Ratio | |
Specific Gravity | |
Work Time | 8 hours |
Comment |