R-2182
LOW COEFFICIENT OF FRICTION SILICONE COATING
Description
  • Two-part silicone elastomer dispersed in xylene
  • Cures to a smooth silicone film that decreases the coefficient of friction (CoF) compared to the uncoated silicone and increases abrasion resistance for moving, sliding, and rubbing parts
  • Provides adhesion to silicone substrates that results in a reduction of surface tack
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • To provide a heat-cured, low-friction coating on cured silicone substrates
  • Apply by spraying for best results (dipping may also be utilized)
Product Details
Property Average Result
Viscosity
Non Volatile Content
Appearance
Cure
Cure System
Mix Ratio
Post Cure 5 minutes / 150 °C
Specific Gravity
Work Time 24 hours
Comment