Description
- Two-part silicone elastomer dispersed in xylene
- Two medium viscosity parts blend easily in a convenient 1:1 ratio (Part A:B)
Applications
- For dip casting and heat-curing of thin elastomeric films
- For use as a coating to minimize ice adhesion
Product Details
Property | Average Result |
---|---|
Viscosity | |
Non Volatile Content | |
Appearance | |
Cure | |
Cure System | |
Durometer | |
Elongation | |
Mix Ratio | |
Refractive Index | |
Stress at Strain | |
Tear | |
Tensile | |
Work Time | 72 hours |
Comment |