R-2160
RED, HIGH TEMPERATURE SILICONE ELASTOMER
Description
  • Two-part, red, pourable silicone system
  • 10:1 Mix Ratio (Part A:B)
  • Cures at room temperature or rapidly with heat
  • High tear strength and good physical properties
Applications
  • As a high temperature adhesive / sealant
  • Ideal for use in potting connectors, cable harness breakouts, molded high voltage terminals, seals and gaskets due to its high physical strength
  • For applications requiring an operating temperature range of -65°C to 315°C (-85°F to 600°F)
Product Details
Property Average Result
Cure System
Viscosity
Durometer
Appearance
Cure
Dielectric Strength
Elongation
Mix Ratio
Non Volatile Content
Operating Temperature
Specific Gravity
Tear
Tensile
Volume Resistivity
Work Time 1.5 hours
Comment