Description
- Two-part, addition-cure black silicone ink dispersed in xylene
- Cures rapidly with heat
Applications
- For use as a marking ink for silicone rubber parts and other components
- Use in applications requiring a rapid cure
- Various application methods including pad printing and screen printing
Product Details
Property | Average Result |
---|---|
Cure System | Platinum |
Viscosity | 900 cP (900 mPa*s) |
Non Volatile Content | 60 % |
Mix Ratio | 1:1 |
Appearance | Black |
Cure | 5 minutes / 150 °C |
Tack Free Time | 24 hours |
Comment | Available in white (R-2100-1), black (R-2100-2), red (R-2100-3), and green (R-2100-6). Dispersed in xylene. |
![](/m-nusil.avantorsciences.com/en_US/images/satellite-in-space-375x235.jpg)
Contact NuSil™ to Order Now
For over 40 years, the NuSil brand has offered a wide range of high-purity silicones. Our team is ready to collaborate with you to provide silicone solutions to satisfy your unique requirements. For additional guidance during your selection process, or to order a sample, contact NuSil today.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.