R-1140
CLEAR SILICONE ADHESIVE
Description
  • One-part, translucent, non-slumping silicone adhesive
  • Supplied ready to use in Semco® cartridges
Applications
  • Bonds aggressively to most surfaces including: glass, ceramics, most metals, natural and synthetic fibers, wood, painted surfaces, many plastics and other silicone rubbers
  • For applications requiring a operating temperature range of -65 to 240°C (-85 to 465°F)
Product Details
Property Average Result
Cure System
Lap Shear 625 psi (4.31 MPa)
Durometer
Appearance
Cure
Elongation
Flow Rate 1.5 in (38.1 mm)
Mix Ratio
Non Volatile Content
Specific Gravity
Tack Free Time
Tear
Tensile
Comment