R-1008-3
RTV SILICONE COATING/MARKING INK
Description
  • One part, flowable silicone elastomer dispersion
  • Cures in 7 days at room temperature when exposed to atmospheric moisture
  • Non-corrosive cure system
  • The R-1008 series is available in Translucent (-0), White (-1), Black (-2), Red (-3), Orange (-4), Yellow (-5), Green (-6), Blue (-7), Violet (-8)
Applications
  • As a marking ink for silicone rubber parts and other components where the coating must maintain long term stability
  • For applications requiring an operating temperature range of -65°C to 260°C
  • (-85°F to 465°F)
Product Details
Property Average Result
Cure System
Viscosity
Non Volatile Content
Mix Ratio
Appearance
Cure
Tack Free Time
Comment