Description
- A two-part, translucent silicone elastomer system that cures at room temperature
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- For applications requiring very rapid cure elastomer
Product Details
Property | Average Result |
---|---|
Durometer | |
Viscosity | |
Work Time | 3 minutes |
Tensile | |
Appearance | |
Cure | |
Cure System | |
Elongation | |
Mix Ratio | |
Specific Gravity | |
Stress at Strain | |
Comment |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.