MED2-4102
COLOR MASTERBATCHES FOR HIGH CONSISTENCY SILICONE ELASTOMERS
Description
- A white, single component masterbatch, with the consistency of a clay like material
- Consists of Barium Sulfate dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
Applications
- For easy and more precise addition of Barium Sulfate to high consistency silicone elastomers, suitable for molding, calendering or extrusion
- Reduces production time and eliminates use of powders that may contaminate clean room environments
Product Details
Property | Average Result |
---|---|
Appearance | |
Mix Ratio | |
Comment |

Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.