Premium Care Line
MED2-4102-6
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A green, single component masterbatch, with the consistency of a clay like material
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
  • Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)
Applications
  • Appearance; Mix Ratio; Plasticity; Extrusion Rate; Specific Gravity
NuSil Technology’s MED2-4102-6 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Appearance
Mix Ratio
Comment
Encapsulation Technology
Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.
Learn more
The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.