Description
- A clear, tacky silicone gel
- A medium viscosity liquid that cures to a soft silicone
- 1:1 Mix Ratio (Part A:B)
Applications
- Ideal for casting, calendering, backfilling and dampening
- Suitable for applications requiring a clear, tacky, silicone gel
- Consider for Transdermal Applications
Product Details
Property | Average Result |
---|---|
Penetration | 15 mm |
Viscosity | |
Mix Ratio | |
Appearance | |
Cure | |
Cure Rate | 3 ppm/°C |
Cure System | |
Non Volatile Content | |
Tack | 6 psi (0.04 MPa) |
Work Time | 2 hours |
Comment |