Premium Care Line
MED-6033
CLEAR LOW VISCOSITY SILICONE ELASTOMER
Description
  • Two-part, pourable clear, silicone elastomer
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property Average Result
Durometer
Viscosity
Work Time 50 hours
Tensile
Appearance
Cure
Cure System
Elongation
Mix Ratio
Refractive Index
Specific Gravity
Tear
Comment