MED-6019
OPTICALLY CLEAR, HIGH DUROMETER LOW VISCOSITY SILICONE ELASTOMER
Description
- Flowable, two-part, optically clear, high durometer silicone
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- Useful in any application requiring a flowable, high durometer material
Product Details
Property | Average Result |
---|---|
Durometer | |
Viscosity | |
Work Time | 2.5 hours |
Tensile | |
Appearance | |
Cure | |
Cure System | |
Elongation | |
Mix Ratio | |
Specific Gravity | |
Tear | |
Comment |