Premium Care Line
MED-6010
OPTICALLY CLEAR LOW VISCOSITY SILICONE ELASTOMER
Description
  • An optically clear, two-part silicone
  • A pourable, medium viscosity material
  • Cures with heat via addition-cure chemistry
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property Average Result
Durometer
Viscosity
Work Time 4 hours
Tensile
Appearance
Cure
Cure System
Elongation
Mix Ratio
Refractive Index
Specific Gravity
Tear
Comment