Description
- A solvent free, two-part, translucent silicone system
- Viscosity and long work time suitable for automatic dispensing
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
- Effective as a dip-coating for encapsulating components
Product Details
Property | Average Result |
---|---|
Durometer | |
Viscosity | |
Work Time | 31 hours |
Tensile | |
Appearance | |
Cure | |
Cure System | |
Elongation | |
Mix Ratio | |
Specific Gravity | |
Comment |