Description
- Two-part, translucent silicone system designed for use with injection molding equipment
- Cures with heat via addition-cure chemistry
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For the injection molding of parts requiring a material with a high durometer including: molded rubber stoppers, gaskets, seals, valves, o-rings and other precision parts
- Suitable for over-molding applications
- Can be used with NuSil’s Healthcare color masterbatches for applications requiring colored silicones
Product Details
Property | Average Result |
---|---|
Durometer | |
Tensile | |
Elongation | |
Tear | |
Appearance | |
Cure | |
Cure System | |
Extrusion Rate | |
Mix Ratio | |
Specific Gravity | |
Stress at Strain | |
Work Time | 72 hours |
Comment |