Implant Line
MED-4719
HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • Two-part silicone elastomer system
  • Cures with heat via addition-cure chemistry
  • Has a non-tacky surface and no volatile by-products or peroxide residues
  • Strained through a 400-mesh screen (minimum)
  • 1:1 Mix Ratio (Part A: Part B)
Applications
  • For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
  • Excellent adhesion and fatigue resistance
NuSil® MED-4719 may be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Durometer
Tensile
Elongation
Tear
Appearance
Cure
Cure System
Cure Type Heat Cure
Mix Ratio
Specific Gravity
Stress at Strain
Work Time 8 hours
Comment