Premium Care Line
MED-4102-5
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A yellow, single component master-batch, with the consistency of a clay like material
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
  • Supported by USP Class VI and ISO 10993 biological testing (reference Biological Testing Data Table)
Applications
  • For easy and more precise pigmentation of high consistency silicone materials, suitable for molding, calendering or extrusion
  • Reduces production time and eliminates use of powders that may contaminate clean room environments
  • Strict quality controls ensure color consistency
NuSil Technology’s MED-4102-5 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Appearance
Mix Ratio
Comment