Premium Care Line
MED-4102-1
COLOR MASTERBATCH FOR HIGH CONSISTENCY SILICONE ELASTOMER
Description
  • A white, single component masterbatch, with the consistency of a clay like material
  • Consists of pigment dispersed in a vinyl-functional silicone polymer which covalently bonds into the matrix of platinum-cured silicone systems
  • Supported by USP Class VI and ISO 10993 Biological testing (reference Biological Testing Data Table)
Applications
  • Appearance; Mix Ratio; Plasticity; Extrusion Rate; Specific Gravity
NuSil Technology’s MED-4102-1 shall not be considered for use in human implantation for a period of greater than 29 days.
Product Details
Property Average Result
Appearance
Mix Ratio
Comment