Description
- Two-part, translucent, pourable silicone elastomer
- Offers high tear strength and excellent physical properties
- Cures with heat via addition-cure chemistry
- 10:1 Mix Ratio (Part A: Part B)
Applications
- For use in potting, encapsulating, coating, and injection or transfer molding applications
Product Details
Property | Average Result |
---|---|
Durometer | |
Viscosity | |
Work Time | 5 hours |
Tensile | |
Appearance | |
Cure | |
Cure System | |
Elongation | |
Mix Ratio | |
Post Cure | 1 minute / 150 °C |
Specific Gravity | |
Stress at Strain | |
Tear | |
Comment |