MED-4019
HIGH CONSISTENCY SILICONE ELASTOMER
Description
- Two-part silicone elastomer system
- Cures with heat via addition-cure chemistry
- Has a non-tacky surface and no volatile by-products or peroxide residues
- Strained through a 400-mesh screen (minimum)
- 1:1 Mix Ratio (Part A: Part B)
Applications
- For a wide variety of fabrication techniques for the healthcare industry including: molding, calendering and extruding
- Excellent adhesion and fatigue resistance
Product Details
Property | Average Result |
---|---|
Durometer | |
Tensile | |
Elongation | |
Tear | |
Appearance | |
Cure | |
Cure System | |
Cure Type | Heat Cure |
Mix Ratio | |
Specific Gravity | |
Stress at Strain | |
Work Time | 5 hours |
Comment |