Description
- Two-part, optically clear, non-phenyl containing addition cure silicone
- Designed to stay thermo-optically stable with heat and UV exposure
- 24 hours minimum work time
- 80 Type A durometer aids with scratch resistance and reduces tackiness
Applications
- For molding or casting hard, non-yellowing lenses for LED, solar or other applications where a hard, optically-stable silicone is needed
- Viscosity is designed to be used with conventional Liquid Injection Molding equipment, and the material is self-leveling for casting application techniques
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Cure | |
Mix Ratio | |
Refractive Index | |
Specific Gravity | |
Tear | |
Work Time | 24 hours |
Comment |