Description
- Two-part, optically clear, silicone system that pours
- Cures at room temperature or rapidly with heat
- Offers good physical properties and a broad operating temperature range
- 1:1 Mix Ratio (Part A:B)
Applications
- For bonding, casting or injection molding of high performance optical components
- For applications requiring index matching at 1.46
Product Details
Property | Average Result |
---|---|
Cure System | |
Viscosity | |
Durometer | |
Tensile | |
Elongation | |
Appearance | |
Cte | |
Cure | |
Dielectric Strength | |
Lap Shear | 510 psi (3.52 MPa) |
Mix Ratio | |
Operating Temperature | |
Refractive Index | |
Specific Gravity | |
Tear | |
Volatile Content | |
Work Time | 2 hours |
Comment |