LS-6941
OPTICAL THERMOSET
Description
  • Two-part, optically clear, solvent free, low viscosity silicone
  • Cures at room temperature or rapidly with heat
  • 10:1 Mix Ratio (Part A:B)
Applications
  • For bonding, casting or injection molding of high performance optical components
  • For applications requiring index matching at 1.41
  • For applications requiring an operating temperature range of -65 to 240°C (–85 to 465°F)
Product Details
Property Average Result
Cure System
Viscosity
Durometer
Tensile
Elongation
Appearance
Cure
Dielectric Strength
Mix Ratio
Operating Temperature
Refractive Index
Specific Gravity
Tack Free Time
Tear
Volume Resistivity
Work Time 5.5 hours
Comment