Description
- A two-part, encapsulation gel
- Extended work time
- 1:1 Mix Ratio (Part A:B)
Applications
- For protection of sensitive photonics assemblies from mechanical shock, thermal shock, dust, and ambient atmosphere
- Extended work time allows voids in complex assemblies to fill in and permits time for any trapped air bubbles to float to the fluid surface and escape
- For applications requiring an operating temperature range of -40°C to 200°C (-40°F to 392°F) or for soldering operations up to 260°C (500°F) for 1 to 2 minutes
Product Details
Property | Average Result |
---|---|
Viscosity | |
Work Time | 11 hours |
Appearance | |
Cure | |
Cure System | |
Durometer | |
Mix Ratio | |
Operating Temperature | |
Refractive Index | |
Specific Gravity | |
Volatile Content | |
Comment |