GEL1-8155

HIGH PURITY DIELECTRIC, FIRM SILICONE GEL

Description

  • High purity, optically clear, firm silicone gel
  • Two medium viscosity components blend easily in a convenient 1:1 ratio by weight or volume

Applications

  • For potting, encapsulating, backfilling, and dampening applications requiring a firm gel with optical clarity
  • Recommended for use with automatic dispense systems

Product Details

Property Average Result
Viscosity
Penetration 0.4 mm
Appearance
Cure
Cure System
Mix Ratio
Non Volatile Content
Operating Temperature
Refractive Index
Comment

Encapsulation Technology

Watch our CareSil team explain the breakthrough Encapsulation Technology and demonstrate how easy it is to create transformative formulations with CES.

Learn more

The test data shown for this material is the average value for typical properties. All of these properties may not be tested on a lot to lot basis and cannot be used to draft specifications. Please contact NuSil for assistance and recommendations in establishing limits for product specifications.