Description
- High purity, optically clear, firm silicone gel
- Two low viscosity components blend easily in a convenient 1:1 ratio by weight or volume
Applications
- For potting, encapsulating, backfilling, and dampening applications requiring a firm gel with optical clarity
- For use in extreme temperatures: features an operating temperature range of -115°C to 265°C (-178°F to 500°F)
Product Details
Property | Average Result |
---|---|
Viscosity | 650 cP (650 mPa*s) |
Penetration | 5 mm |
Work Time | 2 hours |
Appearance | Translucent |
Cure | 30 minutes / 100 °C |
Cure System | Platinum |
Dielectric Strength | 500 volts/mil (19.5 kV/mm) |
Mix Ratio | 1:1 |
Operating Temperature | -115 °C / 265 °C |
Refractive Index | 1.43 |
Specific Gravity | 1 |
Volume Resistivity | 1,000,000,000,000,000 ohmcm |
Comment | Soft Med Tack Gel;For applications requiring a broader operating temperature range. |