Description
- High purity, optically clear, firm silicone gel
- Two components blend easily in a convenient 1:1 ratio by weight or volume
Applications
- For potting, encapsulating, backfilling, and dampening applications requiring a firm gel with optical clarity
- For applications requiring an operating temperature range of -65°C to 200°C (-85°F to 392°F)
Product Details
Property | Average Result |
---|---|
Viscosity | 500 cP (500 mPa*s) |
Penetration | 5 mm |
Work Time | 4 hours |
Appearance | Translucent |
Cure | 30 minutes / 100 °C |
Cure System | Platinum |
Dielectric Strength | 500 volts/mil (19.5 kV/mm) |
Mix Ratio | 1:1 |
Operating Temperature | -65 °C / 200 °C |
Refractive Index | 1.40 |
Specific Gravity | 0.97 |
Volatile Content | 1 % |
Volume Resistivity | 1,000,000,000,000,000 ohmcm |
Comment | Soft High Tack Gel |