GEL-8127
HIGH PURITY DIELECTRIC SILICONE GEL
Description
- Medium to Firm silicone gel
- Low viscosity allows easy mixing and de-airing
- 1:1 Mix Ratio (Part A: Part B)
- Platinum Cure
- RoHS compliant
Applications
- For use as an embedding or potting compound for protection of electronic assemblies and components from environmental contamination
- Provide excellent stress relief for thermal cycling of sensitive components such as wire bonds and thin wafers
- Electrically insulating and non-conductive
- Ideal for potting and filling intricate assemblies
- Ideal for use in automated dispensing equipment
Product Details
Property | Average Result |
---|---|
Viscosity | 530 cP (530 mPa*s) |
Penetration | 8 mm |
Appearance | Translucent |
Cure | 35 minutes / 150 °C |