Description
- Two Part, platinum catalyzed, clear, very soft gel
- 1:1 Mix Ratio (Part A: Part B)
- Cures via addition-cure chemistry
Applications
- For potting, encapsulating, backfilling, and dampening
- Also for applications requiring a low viscosity and low volatility soft gel
Product Details
Property | Average Result |
---|---|
Appearance | |
Cure | |
Cure System | |
Cure Type | Heat Cure |
Mix Ratio | |
Refractive Index | |
Rheology | |
Viscosity | |
Volatile Content |