GEL-3500
FLUOROSILICONE GEL
Description
  • A two-part, firm fluorosilicone gel
  • Available in custom formulations to offer various work times, cure schedules and penetration values to allow matching of processing techniques to production requirements while maintaining the 1:1 mix ratio
Applications
  • For applications requiring a gel with solvent resistance
Product Details
Property Average Result
Viscosity
Work Time 12 hours
Appearance
Cure
Cure System
Durometer
Mix Ratio
Operating Temperature
Comment