Description
- A medium consistency methyl silicone grease
- Typical vapor pressure of 10-8 torr
Applications
- A medium consistency methyl silicone grease
- Typical vapor pressure of 10-8 torr
- For high vacuum and pressure systems
- Specific applications include: glass and ceramic stopcocks, ground glass joints, high temperature gaskets, valves, seals and O-rings
Product Details
Property | Average Result |
---|---|
Viscosity | |
Specific Gravity | |
Volatile Content | |
Appearance | |
Operating Temperature | |
Comment |