FS-3775
FLUOROSILICONE ADHESIVE/SEALANT
Description
  • A translucent, one-part, RTV silicone
  • Based on a fluorosilicone polymer
Applications
  • For sealing and bonding applications requiring solvent and/or fuel resistance
  • Bonds aggressively to most surfaces
  • For protecting or bonding hardware components exposed to gasoline or aviation fuels
  • For applications requiring an operating temperature range of -65°C to 240°C (-85°F to 465°F)
Product Details
Property Average Result
Cure System
Extrusion Rate
Lap Shear 210 psi (1.45 MPa)
Durometer
Appearance
Attribute
Cure
Elongation
Mix Ratio
Non Volatile Content
Specific Gravity
Tack Free Time
Tear
Tensile
Comment