Description
- A white, one-part, fluorosilicone adhesive
- 100 mol% fluorosilicone
- Cures at room temperature
Applications
- For sealing and bonding applications requiring solvent and/or fuel resistance
- Bonds aggressively to most surfaces
- Ideal for protection or bonding of components that are exposed to gasoline or aviation fuels
Product Details
Property | Average Result |
---|---|
Cure System | |
Extrusion Rate | |
Lap Shear | 350 psi (2.41 MPa) |
Durometer | |
Appearance | |
Cure | |
Elongation | |
Mix Ratio | |
Specific Gravity | |
Tack Free Time | |
Tear | |
Tensile | |
Comment |