Description
- A gray, one-part, RTV silicone
- Based on a 100 mole % fluorosilicone
Applications
- For sealing and bonding applications requiring solvent and/or fuel resistance
- Bonds aggressively to most surfaces
- For protecting or bonding hardware components exposed to gasoline or aviation fuels
- For applications requiring an operating temperature range of -85 to 465°F (–65 to 240°C)
Product Details
Property | Average Result |
---|---|
Cure System | |
Lap Shear | 275 psi (1.9 MPa) |
Durometer | |
Appearance | |
Attribute | |
Cure | |
Elongation | |
Mix Ratio | |
Specific Gravity | |
Tack Free Time | |
Tear | |
Tensile | |
Comment |